missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylcinnamaldehyde, predominantly (E), 97%

Catalog No. AAA1042922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1042922 100 g
AAA1042936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1042922 Supplier Thermo Scientific Chemicals Supplier No. A1042922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 101-39-3 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101-39-3
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00006976
InChI Key VLUMOWNVWOXZAU-CLFYSBASSA-N
Synonym alpha-methylcinnamaldehyde, a-methylcinnamaldehyde, 2-propenal, 2-methyl-3-phenyl, 2-methyl-3-phenylprop-2-enal, e-2-methyl-3-phenylpropenal, 2-methyl-3-phenylacrylaldehyde, e-2-methyl-3-phenylacrylaldehyde, 2-methyl-3-phenyl-2-propenal, 2-methyl-3-phenylacrolein, alpha-methyl-trans-cinnamaldehyde
PubChem CID 5372813
SMILES C\C(C=O)=C\C1=CC=CC=C1

Specifications

Density 1.05
Boiling Point 138°C to 140°C (14 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.605
Quantity 100 g
Beilstein 507514
Sensitivity Air Sensitive
IUPAC Name (E)-2-methyl-3-phenylprop-2-enal
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material alpha-Methylcinnamaldehyde, predominantly ( E)
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 202-938-8
RTECSNumber GD6600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only