missing translation for 'onlineSavingsMsg'
Learn More

alpha-Isopropylphenylacetic acid, 97%

CAS: 3508-94-9 | C11H14O2 | 178.231 g/mol

$188.89 - $744.96

Chemical Identifiers

CAS 3508-94-9
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00021708
InChI Key HDLQGISFYDYWFJ-UHFFFAOYSA-N
Synonym alpha-isopropylphenylacetic acid, 2-isopropyl-2-phenylacetic acid, 3-methyl-2-phenyl-butyric acid, 3-methyl-2-phenylbutyric acid, butyric acid, 3-methyl-2-phenyl, benzeneacetic acid, alpha-1-methylethyl, 2-phenylisovaleric acid, acmc-1bn3m, benzeneacetic acid, alpha-1-methylethyl-9ci, alpha-isopropylphenylaceticacid
PubChem CID 19024
IUPAC Name 3-methyl-2-phenylbutanoic acid
SMILES CC(C)C(C1=CC=CC=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1607706
View Documents
Thermo Scientific Chemicals
A1607706
5 g
Each for $188.89
Only null left
 
AAA1607714
View Documents
Thermo Scientific Chemicals
A1607714
25 g
Each for $744.96
Only null left
 
Description

Description

alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

3508-94-9
178.231
HDLQGISFYDYWFJ-UHFFFAOYSA-N
19024
CC(C)C(C1=CC=CC=C1)C(=O)O
C11H14O2
MFCD00021708
alpha-isopropylphenylacetic acid, 2-isopropyl-2-phenylacetic acid, 3-methyl-2-phenyl-butyric acid, 3-methyl-2-phenylbutyric acid, butyric acid, 3-methyl-2-phenyl, benzeneacetic acid, alpha-1-methylethyl, 2-phenylisovaleric acid, acmc-1bn3m, benzeneacetic acid, alpha-1-methylethyl-9ci, alpha-isopropylphenylaceticacid
3-methyl-2-phenylbutanoic acid

Specifications

3508-94-9
C11H14O2
5 g
alpha-isopropylphenylacetic acid, 2-isopropyl-2-phenylacetic acid, 3-methyl-2-phenyl-butyric acid, 3-methyl-2-phenylbutyric acid, butyric acid, 3-methyl-2-phenyl, benzeneacetic acid, alpha-1-methylethyl, 2-phenylisovaleric acid, acmc-1bn3m, benzeneacetic acid, alpha-1-methylethyl-9ci, alpha-isopropylphenylaceticacid
HDLQGISFYDYWFJ-UHFFFAOYSA-N
3-methyl-2-phenylbutanoic acid
19024
97%
57°C to 62°C
MFCD00021708
2045915
Slightly soluble in water.
CC(C)C(C1=CC=CC=C1)C(=O)O
178.231
178.23
alpha-Isopropylphenylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : ET5750060

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only