missing translation for 'onlineSavingsMsg'
Learn More

alpha-Isopropylphenylacetic acid, 97%

Catalog No. aaa1607706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1607706 5 g
AAA1607714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1607706 Supplier Thermo Scientific Chemicals Supplier No. A1607706
Only null left

CAS: 3508-94-9 | C11H14O2 | 178.231 g/mol

alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3508-94-9
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00021708
InChI Key HDLQGISFYDYWFJ-UHFFFAOYSA-N
Synonym alpha-isopropylphenylacetic acid, 2-isopropyl-2-phenylacetic acid, 3-methyl-2-phenyl-butyric acid, 3-methyl-2-phenylbutyric acid, butyric acid, 3-methyl-2-phenyl, benzeneacetic acid, alpha-1-methylethyl, 2-phenylisovaleric acid, acmc-1bn3m, benzeneacetic acid, alpha-1-methylethyl-9ci, alpha-isopropylphenylaceticacid
PubChem CID 19024
IUPAC Name 3-methyl-2-phenylbutanoic acid
SMILES CC(C)C(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 57°C to 62°C
Quantity 5 g
Beilstein 2045915
Solubility Information Slightly soluble in water.
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material alpha-Isopropylphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber ET5750060
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only