missing translation for 'onlineSavingsMsg'
Learn More

alpha-Hexylcinnamaldehyde 90.0+%, TCI America™

Catalog No. H0685500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0685500ML 500 mL
H068525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0685500ML Supplier TCI America Supplier No. H0685500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-86-0
Molecular Formula C15H20O
Molecular Weight (g/mol) 216.32
MDL Number MFCD00006989
InChI Key GUUHFMWKWLOQMM-NTCAYCPXSA-N
Synonym 2-Hexyl-3-phenylacrylaldehyde, 2-Hexyl-3-phenyl-2-propenal
PubChem CID 1715135
IUPAC Name (2E)-2-(phenylmethylidene)octanal
SMILES CCCCCC\C(C=O)=C/C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 175°C
Quantity 500 mL
Formula Weight 216.32
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Hexylcinnamaldehyde
EINECSNumber (3)-2657
RTECSNumber GD6560000
TSCA Yes