missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Galacturonic Acid Hydrate 95.0+%, TCI America™

Catalog No. G00105G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G00105G Supplier TCI America Supplier No. G00105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91510-62-2
Molecular Formula C6H9O7
Molecular Weight (g/mol) 0.00
MDL Number MFCD00006618
InChI Key AEMOLEFTQBMNLQ-DTEWXJGMSA-M
Synonym d-+-galacturonic acid monohydrate
PubChem CID 91872458
IUPAC Name (2S,3R,4S,5R,6R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
SMILES *

Specifications

Quantity 5 g
Formula Weight 194.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-D-Galacturonic Acid Hydrate