missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Cyclodextrin, 98%, Spectrum™ Chemical
Description
Spectrum™ Chemical alpha-Cyclodextrin , a polysaccharide, can be used as a dietary fiber. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 5 kg |
| Packaging | Poly Pail |
| Water | 10% |
| Percent Purity | 98% |
| Grade | Reagent |