missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclodextrin, 97+%

Catalog No. AAJ6068722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6068722 100 g
AAJ6068706 5 g
AAJ6068714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6068722 Supplier Thermo Scientific Chemicals Supplier No. J6068722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful for selective precipitation of enantiomeric, positional or structural isomers | CAS: 10016-20-3 | C36H60O30 | 972.846 g/mol

Useful for selective precipitation of enantiomeric, positional or structural isomersα-Cyclodextrin is used as a fiber ingredient, an odor or flavor masking agent. It is also useful for emulsification applications. It is also used as whipping fiber and emulsifying fiber. It finds application in medical, healthcare and food and beverage applications. It is also used to lower blood low-density lipoprotein cholesterol levels and lower blood triglyceride levels. It plays an essential role in fat free or fat containing dessert compositions and also employed for the reduction or the replacement of egg white in confectionary and bakery applications. Further, it acts as a supramolecular carrier, complexing agent and controlled drug release. In addition to this, it is used to increase the insulin and leptin sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water at 1%(w/v)

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.846
MDL Number MFCD00078207
InChI Key HFHDHCJBZVLPGP-UHFFFAOYSA-N
Synonym cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin
PubChem CID 320760
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O

Specifications

Color White
Quantity 100 g
Merck Index 14,2718
Solubility Information Soluble in water at 1%(w/v)
Optical Rotation +136° (c=10 in Water)
Formula Weight 972.84
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material alpha-Cyclodextrin
EINECSNumber 233-007-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only