missing translation for 'onlineSavingsMsg'
Learn More

alpha-Asarone

Catalog No. AAJ6512506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6512506 5 g
AAJ6512503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6512506 Supplier Thermo Scientific Chemicals Supplier No. J6512506

CAS: 2883-98-9 | C12H16O3 | 208.257 g/mol

α-Asarone was used in the synthesis of series of α-asarone isomers and were investigated for their hypolipidemic and antiplatelet activities. It is used in the synthesis of anti-inflammatory neolignan. Also used in lignan syntheses for antiallergic agents. It is clinically used as medication for treating epilepsy, cough, bronchitis and asthma. It exhibits neuroprotective, anti-oxidative, anticonvulsive and cognitive enhancing action.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform, methanol.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.

Chemical Identifiers

CAS 2883-98-9
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00064457
InChI Key RKFAZBXYICVSKP-AATRIKPKSA-N
Synonym alpha-asarone, asarone, trans-isoasarone, trans-asarone, asaron, azaron, etherophenol, trans-isoasaron, e-asarone, asarum camphor
PubChem CID 636822
ChEBI CHEBI:78309
IUPAC Name 1,2,4-trimethoxy-5-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC(=C(C=C1OC)OC)OC

Specifications

Melting Point 57°C to 61°C
Color White to Yellow
Boiling Point 296°C
Linear Formula (CH3O)3C6H2CH=CHCH3
Quantity 5 g
Beilstein 1910606
Sensitivity Light sensitive
Merck Index 14,824
Solubility Information Soluble in chloroform,methanol.
Formula Weight 208.25
Physical Form Powder
Chemical Name or Material alpha-Asarone
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 220-743-6
RTECSNumber DC2975000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only