missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Difluorophenylacetic acid, 97%

Catalog No. AAH26028MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26028MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH26028MD Supplier Thermo Scientific Chemicals Supplier No. H26028MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 360-03-2 | C8H6F2O2 | 172.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 360-03-2
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00498146
InChI Key PFKSLFZFBCIJOI-UHFFFAOYSA-N
PubChem CID 726153
IUPAC Name 2,2-difluoro-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)(F)F

Specifications

Melting Point 73°C to 76°C
Quantity 250 mg
UN Number UN3261
Formula Weight 172.13
Percent Purity 99%
Chemical Name or Material alpha,alpha-Difluorophenylacetic acid
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 623-610-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only