missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dichloro-p-xylene, 98%

Numéro de catalogue. p-105784
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC147660250 25 g Glass bottle
AC147661000 100 g Glass bottle
AC147665000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC147660250 Fournisseur Thermo Scientific Chemicals Code fournisseur 147660250
Il en reste null

CAS: 623-25-6 | C8H8Cl2 | 175.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 623-25-6
MDL Number MFCD00000920
InChI Key ZZHIDJWUJRKHGX-UHFFFAOYSA-N
Synonym 1,4-bis chloromethyl benzene, alpha,alpha'-dichloro-p-xylene, p-xylylene dichloride, p-xylylene chloride, benzene, 1,4-bis chloromethyl, p-bis chloromethyl benzene, benzene, p-bis chloromethyl, a,a'-dichloro-p-xylene, xylylene dichloride, ccris 1772
PubChem CID 12171
IUPAC Name 1,4-bis(chloromethyl)benzene
SMILES C1=CC(=CC=C1CCl)CCl

Spécifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 98.0°C to 102.0°C
Color White to Yellow
Boiling Point 254.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8Cl2
Linear Formula C6H4(CH2Cl)2
Quantity 25 g
Beilstein 05, 384
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in acetone and methylenechloride,50mg/mL methanol
Molecular Weight (g/mol) 175.06
Formula Weight 175.06
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material α, α'-Dichloro-p-xylene, 98%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 210-782-7
RTECSNumber ZE4100000
TSCA TSCA

RUO – Research Use Only