missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha'-Bis(4-hydroxy-3,5-dimethylphenyl)-1,4-diisopropylbenzene 98.0+%, TCI America™
$504.97 - $504.97
Chemical Identifiers
| CAS | 36395-57-0 |
|---|---|
| Molecular Formula | C28H34O2 |
| Molecular Weight (g/mol) | 402.578 |
| MDL Number | MFCD01321133 |
| InChI Key | SIDYFHJPUCRHJY-UHFFFAOYSA-N |
| PubChem CID | 13652270 |
| IUPAC Name | 4-[2-[4-[2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]phenyl]propan-2-yl]-2,6-dimethylphenol |
| SMILES | CC1=CC(=CC(=C1O)C)C(C)(C)C2=CC=C(C=C2)C(C)(C)C3=CC(=C(C(=C3)C)O)C |
Chemical Identifiers
| 36395-57-0 | |
| 402.578 | |
| SIDYFHJPUCRHJY-UHFFFAOYSA-N | |
| 4-[2-[4-[2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]phenyl]propan-2-yl]-2,6-dimethylphenol |
| C28H34O2 | |
| MFCD01321133 | |
| 13652270 | |
| CC1=CC(=CC(=C1O)C)C(C)(C)C2=CC=C(C=C2)C(C)(C)C3=CC(=C(C(=C3)C)O)C |
Specifications
| 36395-57-0 | |
| Red-White | |
| MFCD01321133 | |
| SIDYFHJPUCRHJY-UHFFFAOYSA-N | |
| 4-[2-[4-[2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]phenyl]propan-2-yl]-2,6-dimethylphenol | |
| 13652270 | |
| ≥98.0% (GC) | |
| alpha,alpha′-Bis(4-hydroxy-3,5-dimethylphenyl)-1,4-diisopropylbenzene |
| 163°C | |
| C28H34O2 | |
| 500 g | |
| CC1=CC(=CC(=C1O)C)C(C)(C)C2=CC=C(C=C2)C(C)(C)C3=CC(=C(C(=C3)C)O)C | |
| 402.578 | |
| 402.58 | |
| Crystalline Powder |
Safety and Handling
TSCA : No