missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Bis(4-hydroxy-3,5-dimethylphenyl)-1,4-diisopropylbenzene 98.0+%, TCI America™

Catalog No. b1850500g
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1850500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1850500G Supplier TCI America Supplier No. B1850500G

Chemical Identifiers

CAS 36395-57-0
Molecular Formula C28H34O2
Molecular Weight (g/mol) 402.578
MDL Number MFCD01321133
InChI Key SIDYFHJPUCRHJY-UHFFFAOYSA-N
PubChem CID 13652270
IUPAC Name 4-[2-[4-[2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]phenyl]propan-2-yl]-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)C(C)(C)C2=CC=C(C=C2)C(C)(C)C3=CC(=C(C(=C3)C)O)C

Specifications

Melting Point 163°C
Color Red-White
Quantity 500 g
Formula Weight 402.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Bis(4-hydroxy-3,5-dimethylphenyl)-1,4-diisopropylbenzene