missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha,alpha',alpha',4,5-Hexabromo-o-xylene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 135°C |
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 1759 |
| Formula Weight | 579.54 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | alpha,alpha,alpha′,alpha′,4,5-Hexabromo-o-xylene |
Safety and Handling
| TSCA | No |