missing translation for 'onlineSavingsMsg'
Learn More

alpha-Acetamidocinnamic Acid 98.0+%, TCI America™

Catalog No. A001425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A001425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A001425G Supplier TCI America Supplier No. A001425G
Only null left

Chemical Identifiers

CAS 5469-45-4
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00008680
InChI Key XODAOBAZOQSFDS-YFHOEESVSA-N
Synonym z-2-acetamido-3-phenylacrylic acid, alpha-acetamidocinnamic acid, .alpha.-acetamidocinnamic acid, .alpha.-acetaminocinnamic acid, acetamidocinnamic acid, z-2-acetamido-3-phenylprop-2-enoic acid, 2-acetylamino-3-phenyl-2-propenoic acid, 2-acetamido-3-phenylprop-2-enoic acid, 2z-2-acetamido-3-phenylprop-2-enoic acid, pubchem8139
PubChem CID 5370579
IUPAC Name (Z)-2-acetamido-3-phenylprop-2-enoic acid
SMILES CC(=O)NC(=CC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 186°C
Color Yellow
Quantity 25 g
Formula Weight 205.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Acetamidocinnamic Acid