Learn More
alpha,4-Dimethylstyrene, stab. with 4-tert-butylcatechol
CAS: 1195-32-0 | C10H12 | 132.21 g/mol
Supplier: Thermo Scientific Chemicals H3173914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| alpha,4-Dimethylstyrene | |
| 1195-32-0 | |
| 186°C to 189°C | |
| C10H12 | |
| MFCD00036510 | |
| 1-methyl-4-prop-1-en-2-yl benzene, dehydro-p-cymene, 2-p-tolylpropene, 4-isopropenyltoluene, 4-methylisopropenylbenzene, 2-p-methylphenyl propene, p-cymenene, p,alpha-dimethylstyrene, p-alpha-dimethylstyrene, 1-isopropenyl-4-methylbenzene | |
| CC(=C)C1=CC=C(C)C=C1 | |
| 132.21 | |
| 132.21 |
| stabilized with 4-tert-butylcatechol | |
| 0.872 | |
| 63°C (145°F) | |
| 1.509 | |
| 25 g | |
| MMSLOZQEMPDGPI-UHFFFAOYSA-N | |
| 1-methyl-4-(prop-1-en-2-yl)benzene | |
| 62385 |
Chemical Identifiers
| 1195-32-0 | |
| 132.21 | |
| MMSLOZQEMPDGPI-UHFFFAOYSA-N | |
| 62385 |
| C10H12 | |
| MFCD00036510 | |
| 1-methyl-4-prop-1-en-2-yl benzene, dehydro-p-cymene, 2-p-tolylpropene, 4-isopropenyltoluene, 4-methylisopropenylbenzene, 2-p-methylphenyl propene, p-cymenene, p,alpha-dimethylstyrene, p-alpha-dimethylstyrene, 1-isopropenyl-4-methylbenzene | |
| CC(=C)C1=CC=C(C)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 214-795-9
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only