accessibility menu, dialog, popup

UserName

H3CCH2CH3

CAS RN 1195-32-0

IUPAC Name: 1-methyl-4-(prop-1-en-2-yl)benzene
Synonyms: P,α-dimethylstyrene
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: MMSLOZQEMPDGPI-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC=C(C)C=C1
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)