missing translation for 'onlineSavingsMsg'
Learn More

alpha,4-Dimethylstyrene, stab. with 4-tert-butylcatechol

Catalog No. p-7030859
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3173906 5 g
AAH3173914 25 g
AAH3173922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3173906 Supplier Thermo Scientific Chemicals Supplier No. H3173906
Only null left

CAS: 1195-32-0 | C10H12 | 132.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1195-32-0
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00036510
InChI Key MMSLOZQEMPDGPI-UHFFFAOYSA-N
Synonym 1-methyl-4-prop-1-en-2-yl benzene, dehydro-p-cymene, 2-p-tolylpropene, 4-isopropenyltoluene, 4-methylisopropenylbenzene, 2-p-methylphenyl propene, p-cymenene, p,alpha-dimethylstyrene, p-alpha-dimethylstyrene, 1-isopropenyl-4-methylbenzene
PubChem CID 62385
SMILES CC(=C)C1=CC=C(C)C=C1

Specifications

Density 0.872
Boiling Point 186°C to 189°C
Flash Point 63°C (145°F)
Refractive Index 1.509
Quantity 5 g
IUPAC Name 1-methyl-4-prop-1-en-2-ylbenzene
Formula Weight 132.21
Chemical Name or Material alpha,4-Dimethylstyrene, stabilized with 4-tert-butylcatechol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 214-795-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only