missing translation for 'onlineSavingsMsg'
Learn More

alpha,3,5-Tribromo-2-hydroxytoluene 98.0+%, TCI America™

Catalog No. d017225g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D017225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D017225G Supplier TCI America Supplier No. D017225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4186-54-3
Molecular Formula C7H5Br3O
Molecular Weight (g/mol) 344.828
MDL Number MFCD00059612
InChI Key SPUZPJKEJXIRSF-UHFFFAOYSA-N
Synonym 2,4-Dibromo-6-bromomethylphenol, 3,5-Dibromo-2-hydroxybenzyl Bromide
PubChem CID 622177
IUPAC Name 2,4-dibromo-6-(bromomethyl)phenol
SMILES C1=C(C=C(C(=C1Br)O)CBr)Br

Specifications

Melting Point 118°C
Quantity 25 g
Formula Weight 344.83
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha,3,5-Tribromo-2-hydroxytoluene