missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha-1 Microglobulin Recombinant Rabbit Monoclonal Antibody (23GB3200)

Rabbit Recombinant Monoclonal Antibody
Spécifications
| Antigen | alpha-1 Microglobulin |
|---|---|
| Clone | 23GB3200 |
| Concentration | 0.45 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
MA553604
![]() |
Invitrogen™
MA553604 |
100 μL |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
MA553605
|
Invitrogen™
MA553605 |
20 μL |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
alpha-1 Microglobulin Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes a complex glycoprotein secreted in plasma. The precursor is proteolytically processed into distinct functioning proteins: alpha-1-microglobulin, which belongs to the superfamily of lipocalin transport proteins and may play a role in the regulation of inflammatory processes, and bikunin, which is a urinary trypsin inhibitor belonging to the superfamily of Kunitz-type protease inhibitors and plays an important role in many physiological and pathological processes. This gene is located on chromosome 9 in a cluster of lipocalin genes.Spécifications
| alpha-1 Microglobulin | |
| 0.45 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P02760 | |
| 259 | |
| A synthetic peptide of human AMBP (50-100AA). | |
| Antibody |
| 23GB3200 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| AMBP | |
| A1M; AI194774; alpha 1 microglobulin/bikunin; alpha-1 microglobulin; Alpha-1 microglobulin/bikunin; Alpha-1 microglycoprotein; Alpha-1-microglobulin; alpha-1-microglobulin/bikunin; alpha1-microglobulin/bikunin; alpha-1-microglobulin/bikunin precursor; alpha-1-microglobulin-bikunin; alpha1-microglobulin-bikunin precursor; Ambp; AMBP protein; ASPI; BI-14; Bikunin; complex-forming glycoprotein heterogeneous in charge; Cumulus extracellular matrix-stabilizing factor; EDC1; ESF; growth-inhibiting protein 19; HCP; HI30; HI-30; IATIL; inter-alpha-trypsin inhibitor (protein HC), light; inter-alpha-trypsin inhibitor light chain; Intin4; ITI; ITIL; ITILC; ITI-LC; polyprotein; protein AMBP; Protein HC; Protein Protein HC; trypstatin; ulinastatin; unnamed protein product; Urinary Trypsin Inhibitor; uristatin; uronic-acid-rich protein; UTI | |
| AMBP | |
| Primary | |
| Affinity chromatography |