missing translation for 'onlineSavingsMsg'
Learn More

Aloe Emodin 97.0+%, TCI America™

Catalog No. A24911G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24911G 1 g
A2491100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24911G Supplier TCI America Supplier No. A24911G

Chemical Identifiers

CAS 481-72-1
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00017373
InChI Key YDQWDHRMZQUTBA-UHFFFAOYSA-N
Synonym 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone
PubChem CID 10207
ChEBI CHEBI:2607
IUPAC Name 1,8-dihydroxy-3-(hydroxymethyl)anthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)CO)O

Specifications

Melting Point 222°C
Quantity 1 g
Formula Weight 270.24
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Aloe Emodin
RTECSNumber CB6712200
TSCA No
Recommended Storage Refrigerator