missing translation for 'onlineSavingsMsg'
Learn More

Allyltriisopropylsilane 97.0+%, TCI America™

Catalog No. A18585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18585G Supplier TCI America Supplier No. A18585G
Only null left

Chemical Identifiers

CAS 24400-84-8
Molecular Formula C12H26Si
Molecular Weight (g/mol) 198.425
InChI Key AKQHUJRZKBYZLC-UHFFFAOYSA-N
PubChem CID 5018111
IUPAC Name tri(propan-2-yl)-prop-2-enylsilane
SMILES CC(C)[Si](CC=C)(C(C)C)C(C)C

Specifications

Color Colorless
Quantity 5 g
Formula Weight 198.43
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyltriisopropylsilane