missing translation for 'onlineSavingsMsg'
Learn More

Allyloxytrimethylsilane 97.0+%, TCI America™

Catalog No. A092725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A092725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A092725ML Supplier TCI America Supplier No. A092725ML

Chemical Identifiers

CAS 18146-00-4
Molecular Formula C6H14OSi
Molecular Weight (g/mol) 130.262
MDL Number MFCD00008643
InChI Key MNMVKGDEKPPREK-UHFFFAOYSA-N
Synonym allyloxytrimethylsilane, allyl trimethylsilyl ether, silane, trimethyl 2-propenyloxy, trimethyl 2-propenyloxy silane, o-trimethylsilylallyl alcohol, trimethylallyloxysilane, allyloxy trimethylsilane, silane, trimethyl 2-propen-1-yloxy, trimethyl prop-2-en-1-yloxy silane, acmc-1boq9
PubChem CID 87471
IUPAC Name trimethyl(prop-2-enoxy)silane
SMILES C[Si](C)(C)OCC=C

Specifications

Color Colorless
Boiling Point 100°C
Quantity 25 mL
UN Number 1993
Formula Weight 130.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyloxytrimethylsilane
EINECSNumber (2)-2960
TSCA Yes