missing translation for 'onlineSavingsMsg'
Learn More

Allyl Trifluoroacetate 95.0+%, TCI America™

Catalog No. T17655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17655G 5 g
T176525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17655G Supplier TCI America Supplier No. T17655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 383-67-5
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.088
MDL Number MFCD00013567
InChI Key XIVPVSIDXBTZLM-UHFFFAOYSA-N
Synonym allyl trifluoroacetate, allyl trifluoracetate, trifluoroacetic acid allyl ester, prop-2-en-1-yl 2,2,2-trifluoroacetate, allyltrifluoroacetate, pubchem12548, acmc-209izu, allyl alcohol, trifluoroacetate, acetic acid, trifluoro-, allyl ester
PubChem CID 67845
IUPAC Name prop-2-enyl 2,2,2-trifluoroacetate
SMILES C=CCOC(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 66°C
Quantity 5 g
UN Number 2924
Formula Weight 154.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Trifluoroacetate