missing translation for 'onlineSavingsMsg'
Learn More

Allyl Sulfide 98.0+%, TCI America™

Catalog No. A023525ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A023525ML 25 mL
A0235250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A023525ML Supplier TCI America Supplier No. A023525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 592-88-1
Molecular Formula C6H10S
Molecular Weight (g/mol) 114.21
MDL Number MFCD00008658
InChI Key UBJVUCKUDDKUJF-UHFFFAOYSA-N
Synonym diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide
PubChem CID 11617
ChEBI CHEBI:4489
IUPAC Name 3-(prop-2-en-1-ylsulfanyl)prop-1-ene
SMILES C=CCSCC=C

Specifications

Color Colorless
Boiling Point 138°C
Quantity 25 mL
UN Number 1993
Formula Weight 114.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Sulfide
EINECSNumber (2)-0471
RTECSNumber BC4900000
TSCA Yes