missing translation for 'onlineSavingsMsg'
Learn More

Allyl Propyl Sulfide 97.0+%, TCI America™

Catalog No. A10615ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10615ML 5 mL
A106125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10615ML Supplier TCI America Supplier No. A10615ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27817-67-0
Molecular Formula C6H12S
Molecular Weight (g/mol) 116.222
MDL Number MFCD00015220
InChI Key JMLIYQJQKZYHDQ-UHFFFAOYSA-N
PubChem CID 98781
IUPAC Name 1-prop-2-enylsulfanylpropane
SMILES CCCSCC=C

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 mL
UN Number 1993
Formula Weight 116.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Propyl Sulfide