missing translation for 'onlineSavingsMsg'
Learn More

Allyl Phenyl Sulfide 98.0+%, TCI America™

Catalog No. A14425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14425G 5 g
A144225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14425G Supplier TCI America Supplier No. A14425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5296-64-0
Molecular Formula C9H10S
Molecular Weight (g/mol) 150.24
MDL Number MFCD00014957
InChI Key QGNRLAFFKKBSIM-UHFFFAOYSA-N
Synonym allyl phenyl sulfide, allylthio benzene, allyl phenyl sulfane, benzene, 2-propenylthio, allylphenyl sulfide, phenyl allyl sulfide, allylphenylthioether, allyl phenyl ;sulfane, sulfide, allyl phenyl
PubChem CID 79180
IUPAC Name (prop-2-en-1-ylsulfanyl)benzene
SMILES C=CCSC1=CC=CC=C1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 150.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Phenyl Sulfide