missing translation for 'onlineSavingsMsg'
Learn More

Allyl Glycidyl Ether, 99+%

Catalog No. AC102945000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102945000 500 g Glass bottle
AC102941000 100 g Glass bottle
AC102940025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102945000 Supplier Thermo Scientific Chemicals Supplier No. 102945000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-92-3 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless
Density 0.9620g/mL
Boiling Point 154.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4332 to 1.4352
Quantity 500 g
Specific Gravity 0.962
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether and,benzene
Formula Weight 114.14
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Allyl glycidyl ether, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Harmful if swallowed.
May cause respiratory irri
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 203-442-4
RTECSNumber RR0875000
TSCA TSCA

RUO – Research Use Only