missing translation for 'onlineSavingsMsg'
Learn More

Allyl glycidyl ether, 99+%, AcroSeal™

Catalog No. AC430958000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430958000 800 mL AcroSeal™ Glass Bottle
AC430951000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430958000 Supplier Thermo Scientific Chemicals Supplier No. 430958000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-92-3 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless
Density 0.9620g/mL
Boiling Point 154.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4332 to 1.4352
Quantity 800 mL
Specific Gravity 0.962
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether and,benzene
Formula Weight 114.14
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Allyl glycidyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Harmful if swallowed.
May cause respiratory irri
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 203-442-4

RUO – Research Use Only