missing translation for 'onlineSavingsMsg'
Learn More

Allyl ethyl sulfide, 97%

Catalog No. AAL1284706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1284706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1284706 Supplier Thermo Scientific Chemicals Supplier No. L1284706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5296-62-8 | C5H10S | 102.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5296-62-8
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00026979
InChI Key NOJXPGXFDASWEI-UHFFFAOYSA-N
Synonym allyl ethyl sulfide, 4-thia-1-hexene, 1-propene-1-one, 3-ethylthio, allylethylsulfide, allyl ethylsulfide, allylethyl sulfide, allyl ethyl sulphide, 3-ethylthio-1-propene, acmc-1ao1o
PubChem CID 123229
IUPAC Name 3-ethylsulfanylprop-1-ene
SMILES CCSCC=C

Specifications

Density 0.868
Boiling Point 115°C to 116°C
Flash Point 18°C (64°F)
Odor Unpleasant
Refractive Index 1.468
Linear Formula CH3CH2SCH2CH=CH2
Quantity 5 g
UN Number UN1993
Beilstein 1736876
Formula Weight 102.19
Percent Purity 97%
Chemical Name or Material Allyl ethyl sulfide
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only