missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl cyanide, 97%
Description
Allyl cyanide is used as an anti-microbial. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water (3 g/L) at 20°C.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Soluble in water (3 g/L) at 20°C.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Spécifications
Spécifications
| Melting Point | -87°C |
| Color | Brown to Colorless |
| Density | 0.834 g/mL |
| Boiling Point | 118°C to 120°C |
| Flash Point | 24°C (75°F) |
| Odor | Unpleasant |
| Refractive Index | 1.405 |
| Linear Formula | CH2=CHCH2CN |
| Quantity | 25 g |
| UN Number | UN1992 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H226-H301-H312-H318-H330-H360D-H373 |
| Hazard Statement | GHS H Statement H301-H331-H226-H361-H312-H319-H335 Toxic if swallowed. Toxic if inhaled. Flammable liquid and vapor. Suspected of damaging fertility or the unborn child. Harmful in contact with skin. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S. |
| EINECSNumber | 203-701-1 |
| RTECSNumber | EM8050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only