missing translation for 'onlineSavingsMsg'
Learn More

Allyl Chloride 98.0+%, TCI America™

Catalog No. C027425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C027425ML 25 mL
C0274500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C027425ML Supplier TCI America Supplier No. C027425ML

Chemical Identifiers

CAS 107-05-1
Molecular Formula C3H5Cl
Molecular Weight (g/mol) 76.523
MDL Number MFCD00000984
InChI Key OSDWBNJEKMUWAV-UHFFFAOYSA-N
Synonym allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
PubChem CID 7850
ChEBI CHEBI:82379
IUPAC Name 3-chloroprop-1-ene
SMILES C=CCCl

Specifications

Color Yellow
Boiling Point 45°C
Quantity 25 mL
UN Number 1100
Formula Weight 76.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Chloride
EINECSNumber (2)-0123
RTECSNumber UC7350000
TSCA Yes
Recommended Storage Refrigerator