missing translation for 'onlineSavingsMsg'
Learn More

Allopurinol 98.0+%, TCI America™

Catalog No. A090725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A090725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A090725G Supplier TCI America Supplier No. A090725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 315-30-0
Melting Point 384°C
Color White-Yellow
Molecular Formula C5H4N4O
Quantity 25 g
MDL Number MFCD00599413
UN Number 2811
Synonym allopurinol, 1h-pyrazolo 3,4-d pyrimidin-4-ol, zyloprim, zyloric, lopurin, atisuril, bleminol, caplenal, uripurinol, embarin
InChI Key OFCNXPDARWKPPY-UHFFFAOYSA-N
SMILES O=C1N=CN=C2NNC=C12
IUPAC Name 1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
Molecular Weight (g/mol) 136.11
PubChem CID 2094
ChEBI CHEBI:40279
Formula Weight 136.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Allopurinol
Show More Show Less
RTECSNumber UR0785000
TSCA Yes