missing translation for 'onlineSavingsMsg'
Learn More

Allopurinol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019565
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019565 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019565 Supplier Merck Emd Millipore Supplier No. PHR13771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Allopurinol is a drug used in the treatment of chronic gout or hyperuricaemia. Its mode of action involves inhibition of xanthine oxidase, an important enzyme catalyzing the hydroxylation of hypoxanthine to xanthine and xanthine to uric acid. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 315-30-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C5H4N4 O
MDL Number MFCD00599413
Melting Point >300°C
Synonym Allopurinol; 1 H-Pyrazolo(3,4-d)pyrimidin-4-ol; 4-Hydroxypyrazolo(3,4-d)pyrimidine; 4-Hydroxypyrazolo[3,4-d]pyrimidine; HPP
Type Certified Reference Material
Quantity 1 g
Formula Weight 136.11
Grade Certified Reference Material
Show More Show Less