missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allopurinol Impurity B, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 22407-20-1 |
| Molecular Formula | C5H6N4O2 |
| Linear Formula | C5H6N4O2 |
| Synonym | 3-(Formylamino)-1 H-pyrazole-4-carboxamide; 5-(Formylamino)-1 H-pyrazole-4-carboxamide |
| Quantity | 10 mg |
| Formula Weight | 154.13 |
| Grade | British Pharmacopoeia (BP) Reference Standard |