missing translation for 'onlineSavingsMsg'
Learn More

Allantoin 98.0+%, TCI America™

Catalog No. A021125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A021125G 25 g
A0211500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A021125G Supplier TCI America Supplier No. A021125G

Chemical Identifiers

CAS 97-59-6
Molecular Formula C4H6N4O3
Molecular Weight (g/mol) 158.117
MDL Number MFCD00005260
InChI Key POJWUDADGALRAB-UHFFFAOYSA-N
Synonym allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream
PubChem CID 204
ChEBI CHEBI:15676
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
SMILES C1(C(=O)NC(=O)N1)NC(=O)N

Specifications

Melting Point 223°C
Color White
Quantity 25 g
Formula Weight 158.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Allantoin
EINECSNumber (9)-0199
RTECSNumber YT1600000
TSCA Yes