Learn More
Cytiva AlkPhos Direct™ Hybridization Buffer for Direct Labeling System
Description
Includes
Each system includes the following reagents, sufficient for 25 or 50 labeling reactions and detection of 5000cm2 membrane: labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star™ or ECF detection reagents.
- Hybridization stringency can be controlled by elevating temperatures up to 75°C (167°F), or by varying salt concentration
- Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
- ECF™ detection for quantitation experiments
- Thirty minutes from completion of hybridization to application of detection reagent: saves two to three hours over conventional indirect methods
- Resulting probes can be used without purification
Specifications
Specifications
| For Use With (Application) | Fast, convenient for high-sensitivity applications with DNA, RNA, Oligo probes |
| Includes | Labeling reagent, crosslinker, control DNA, Reaction Buffer, hybridization Buffer, blocking agent, CDP-Star™ or ECF detection reagents |
| Product Type | Amersham AlkPhos Direct Hybridization Buffer sufficient for 5000 cm2 |
| Quantity | For 5000 cm2 membrane |