missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Titration Solvent, Ricca Chemical

Catalog No. RSOD118020F
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOD118020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOD118020F Supplier Ricca Chemical Company Supplier No. RSOD118020F

Specifications

Chemical Name or Material Alkaline Titration Solvent
Color Colorless
Name Note Sodium Acetate Trihydrate (<5%), Isopropyl Alcohol, Water
CAS 6131-90-4, 67-63-0, 7732-18-5
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Characteristic alcohol odor
Packaging Cubitainer
Solvent or Matrix Nonaqueous Solution
Quantity 20 L
Physical Form Liquid
Show More Show Less