missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Pyrogallol Reagent, Orsat, Ricca Chemical

Catalog No. 55716
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
55716 500 mL Amber Poly Bottle
55732 1 L Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55716 Supplier Ricca Chemical Company Supplier No. RI55716

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-66-1 , 1310-58-3 , 7732-18-5
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

CAS Min % 4.0, 25.0, 65.0
CAS Max % 6.0, 35.0, 75.0
Packaging Amber Poly Bottle
Quantity 500 mL
Chemical Name or Material Alkaline Pyrogallol Reagent