Learn More
Bioassay Systems QuantiChrom™ Alkaline Phosphatase Assay Kit. For quantitative determination of alkaline phosphatase (ALP) activity using stable p-nitrophenol phosphate substrate.
Numéro de catalogue. 50489198
Encompass
For quantitative determination of alkaline phosphatase (ALP) activity using stable p-nitrophenol phosphate substrate. Key Features: High sensitivity and wide linear range. Use 5 µL serum or plasma sample. The detection limit is 2 U/L, linear up to 800 U/L. Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of ALP activity within 5 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: OD405nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 4 min. Kit size: 250 tests. Detection limit: 2 U/L.
Numéro de catalogue. 50489198 Fournisseur Bioassay Systems Code fournisseur DALP250
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
For quantitative determination of alkaline phosphatase (ALP) activity using stable p-nitrophenol phosphate substrate. Key Features: High sensitivity and wide linear range. Use 5 µL serum or plasma sample. The detection limit is 2 U/L, linear up to 800 U/L. Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of ALP activity within 5 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: OD405nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 4 min. Kit size: 250 tests. Detection limit: 2 U/L.