Learn More
SignalChem Biotech ABL1 (K290R) Protein 1000ug
Numéro de catalogue. 502576066
Encompass
Recombinant human ABL1 (K290R) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. ABL1 (K290R) is a kinase dead mutant.
Numéro de catalogue. 502576066 Fournisseur SignalChem Biotech Code fournisseur A0316NG1000
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant human ABL1 (K290R) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. ABL1 (K290R) is a kinase dead mutant.