missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizarin Yellow R sodium salt
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown to Yellow |
| Physical Form | Powder |
| Quantity | 25 g |
| Beilstein | 962365 |
| Formula Weight | 309.22 |
| Odor | Odorless |
| Appearance | Dark yellow to brown powder |
| Chemical Name or Material | Alizarin Yellow R sodium salt |
Safety and Handling
| Hazard Category | H302 |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 217-002-4 |
| RTECSNumber | DH2528600 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only