missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Yellow R sodium salt

Catalog No. AA3870709
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3870709 10 g
AA3870714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3870709 Supplier Thermo Scientific Chemicals Supplier No. 03870709
Only null left

CAS: 1718-34-9 | C13H8N3NaO5 | 309.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1718-34-9
Molecular Formula C13H8N3NaO5
Molecular Weight (g/mol) 309.213
MDL Number MFCD00067121
InChI Key HKSANXZSKDUDBD-UHFFFAOYSA-M
Synonym alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt,
PubChem CID 44135758
IUPAC Name sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]

Specifications

Color Brown to Yellow
Physical Form Powder
Quantity 10 g
Beilstein 962365
Formula Weight 309.22
Odor Odorless
Appearance Dark yellow to brown powder
Chemical Name or Material Alizarin Yellow R sodium salt
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 217-002-4
RTECSNumber DH2528600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only