missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alizarin Red S, pure, certified

Catalog No. AC400480250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC400480250 25 g
AC400481000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400480250 Supplier Thermo Scientific Chemicals Supplier No. 40048 0250

CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
MDL Number MFCD00013049
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
Synonym 9,10-Dihydro-3,4-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid, sodium salt, 9, 10-Dihydro-3, 4-dihydroxy-9, Mordant Red 3, Sodium alizarinesulfonate
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

Absorption Ratio 0.97 to 1.07 (for the first maximum) (P-15/P+15)
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in ethanol
Infrared Spectrum Authentic
Lambda Max 554 to 558nm 590 to 599nm
Formula Weight 342.26
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 3.5 (yellow) to pH 6.5 (pink) and from pH 9.4 (orange-brown) to pH 12.0 (violet)
Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Alizarin Red S
Show More Show Less
Hazard Category Warning
EINECSNumber 204-981-8
RTECSNumber CB1095300
TSCA TSCA

RUO – Research Use Only