missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alizarin Complexone dihydrate, pure, indicator grade

Catalog No. AC155830050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC155830050 5 g
AC155830010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155830050 Supplier Thermo Scientific Chemicals Supplier No. 155830050

CAS: 455303-00-1 | C19H19NO10 | 421.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 455303-00-1
Molecular Formula C19H19NO10
Molecular Weight (g/mol) 421.36
MDL Number MFCD00149067
InChI Key LLNGUADXYFYGGJ-UHFFFAOYSA-N
Synonym (3, 4-Dihydroxy-2-anthraquinonyl)methyliminodiacetic acid dihydrate, Alizarin fluorine blue
PubChem CID 23624113
SMILES O.O.OC(=O)CN(CC(O)=O)CC1=CC2=C(C(O)=C1O)C(=O)C1=C(C=CC=C1)C2=O

Specifications

Absorbance 0.42 min. (lambda max, after 1 hr.) (2mL of a 0.2% soln. in 0.02M NaOH + 5mL acetate buffer (pH 5.1) diluted with H2O to 100mL), (1cm cell vs. water)
Melting Point 180.0°C to 185.0°C
Infrared Spectrum Authentic
Lambda Max 425 to 430nm
Packaging Glass bottle
Color Brown to Yellow
Solubility Information Solubility in water: slightly soluble (0.2g/L). Other solubilities: soluble in aqueous alkali, insoluble in alcohol and ether
Quantity 5 g
Physical Form Powder
IUPAC Name 2-[(carboxymethyl)[(3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl]amino]acetic acid dihydrate
Formula Weight 421.35
Percent Purity 97 to 103% (on dry substance)
Grade Pure
Chemical Name or Material Alizarin Complexone dihydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-544-2
RTECSNumber AH0585000
TSCA TSCA

RUO – Research Use Only