missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Alginic acid, sodium salt
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 250 g |
| Packaging | Plastic Bottle |
| Chemical Name or Material | Alginic acid, sodium salt |
Frequently Asked Questions (FAQs)
What is the molecular weight and M/G ratio of Alginic acid, sodium salt (Cat. No. 177775000, 177770050, 177772500, 177770025)?
The molecular weight of Alginic acid, sodium salt (Cat. No. 177775000, 177770050, 177772500, 177770025) is 450,000-550,000 Dalton and the M/G ratio is 1.5-1.6, thus ~60% mannuronic acid (m) and ~40% guluronic acid (n).
Safety and Handling
| RTECSNumber | AZ5820000 |
| TSCA | TSCA |
RUO – Research Use Only