missing translation for 'onlineSavingsMsg'
Learn More

Alfuzosin Hydrochloride 98.0+%, TCI America™

Catalog No. A25911G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25911G 1 g
A2591100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25911G Supplier TCI America Supplier No. A25911G
Only null left

Chemical Identifiers

CAS 81403-68-1
Molecular Formula C19H28ClN5O4
Molecular Weight (g/mol) 425.914
MDL Number MFCD00879135
InChI Key YTNKWDJILNVLGX-UHFFFAOYSA-N
Synonym alfuzosin hydrochloride, uroxatral, alfuzosin hcl, xatral, urion, alfoten, alfetim, xatral retard, xatral od
PubChem CID 71764
ChEBI CHEBI:32286
IUPAC Name N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide;hydrochloride
SMILES CN(CCCNC(=O)C1CCCO1)C2=NC3=CC(=C(C=C3C(=N2)N)OC)OC.Cl

Specifications

Melting Point 229°C
Color White
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 425.91
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Alfuzosin Hydrochloride
RTECSNumber LT9965475
TSCA No