Learn More
Invitrogen™ Alexa Fluor™ 647-Poly-GT Peptide Substrate
A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.
$580.00 - $1,670.00
Specifications
| Color | Far-Red |
|---|---|
| Detection Method | Fluorescence |
| Form | Liquid |
| Substrate | Random (Glu:Tyr) polymer in a ratio of 4:1 |
| Substrate Properties | Peptide-Based Substrate |
Description
A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.
For use in combination with the LanthaScreen™ Eu-PY20 Antibody (Cat. No. PV5696) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
| Far-Red | |
| Liquid | |
| Peptide-Based Substrate | |
| Tyrosine Kinases | |
| Alexa Fluor™ 647 | |
| TR-FRET |
| Fluorescence | |
| Random (Glu:Tyr) polymer in a ratio of 4:1 | |
| Kinase Substrate | |
| 340/665 nm | |
| Dry Ice | |
| EGFR |
For Research Use Only. Not for use in diagnostic procedures.