missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 647-Poly-GT Peptide Substrate

Catalog No. PV5693
Encompass
Change view
Click to view available options
Quantity:
1 nmol
10 nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5693 1 nmol
PV5836 10 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV5693 Supplier Invitrogen™ Supplier No. PV5693

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.

For use in combination with the LanthaScreen™ Eu-PY20 Antibody (Cat. No. PV5696) to evaluate inhibitors in medium to high throughput screening applications.

Specifications

Assay Entry Biochemical europium phospho-specific activity
Color Far-Red
Content And Storage 30 μM AlexaFluor 647 Poly-GT peptide substrate in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl, pH 7.5 (HEPES buffered saline).

Store at -20°C and protect from light.
Detection Method Fluorescence
Form Liquid
Product Line LanthaScreen
Quantity 1 nmol
Substrate Random (Glu:Tyr) polymer in a ratio of 4:1
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Tyrosine Kinases
Excitation/Emission 340/665 nm
Label or Dye Alexa Fluor™ 647
Shipping Condition Dry Ice
Target Entry ABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA V561D, SYK, TYRO3, ZAP70, FGFR1, FGFR2, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, TEK, FLT1, MUSK, SRCN1, ABL1 E255K, EPHA4, FYN, PTK2, PTK2B, RET, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, ABL1, ABL2, AXL, FGFR3, FGR, HCK, KIT V654A, EPHA1, EPHA8, EPHB3, INSRR, LYNB, NTRK1, PDGFRA D842V, SRMS, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, EPHA7, EPHB1, FRK, ITK, FLT3 D835Y, SRC, BTK, FES, ERBB2, NTRK2
Technique TR-FRET
Validated Kinases EGFR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.