missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Aldrich™ TLC Developing Tank - Cylindrical Glass Tank

Catalog No. 111010863
Change view
Click to view available options
Height (Metric):
10.5 cm
21 cm
Quantity:
1/Pk.
6 x 1/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Quantity
111010863 10.5 cm 1/Pk.
111008839 10.5 cm 6 x 1/Pk.
111010723 21 cm 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111010863 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2439061EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aldrich™ TLC developing tanks are used to develop TLC plates.

Can be used as a staining chamber. Made of glass with lid. Compact size offers good plate visibility and minimal solvent use.

Specifications

Material Glass
Quantity 1/Pk.
For Use With (Application) TLC
Product Line Aldrich
Height (Metric) 10.5 cm
Diameter (Metric) Outer 6.5 cm

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC