missing translation for 'onlineSavingsMsg'
Learn More

Albumin, Egg, Powder, Purified, J.T. Baker™

Numéro de catalogue. 2002211
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
2002211 125 g Poly Bottle
2002212 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 2002211 Fournisseur Avantor Code fournisseur 044004
Il en reste null

Identifiants chimiques

CAS 9006-50-2
Molecular Formula C6H8N2O4
Molecular Weight (g/mol) 172.14
InChI Key QCVGEOXPDFCNHA-UHFFFAOYSA-N
Synonym tridodeoylamine
PubChem CID 25022239
IUPAC Name 5,5-dimethyl-2,4-dioxo-1,3-oxazolidine-3-carboxamide
SMILES CC1(C(=O)N(C(=O)O1)C(=O)N)C

Spécifications

Packaging Poly Bottle
Quantity 125 g
Grade Purified
Physical Form Powder
Chemical Name or Material Albumin, Egg